missing translation for 'onlineSavingsMsg'
Learn More

Trichlorosilane, 98%

Product Code. 15451057
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15451057

Brand: Thermo Scientific Alfa Aesar 014078.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 10025-78-2 | Cl3Si | 134.435 g/mol

Trichlorosilane is used to prepare silicon for semiconductors. It is an active ingredient used in the preparation of polysilicons and silicone resins. It acts as coupling agents and silane adhesion promoter. It is a precursor to prepare organosilicon compounds through hydrosilylation reactions. Further, it is involved in the preparation of octadecyltrichlorosilane, perfluoroctyltrichlorosilane, and perfluordecyltrichlorosilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-78-2
Molecular Formula Cl3Si
Molecular Weight (g/mol) 134.435
MDL Number MFCD00011518
InChI Key PPDADIYYMSXQJK-UHFFFAOYSA-N
Synonym trichlorosilane, silane, trichloro, silicochloroform, trichloorsilaan, trichlorosilyl, trichloromonosilane, silane a-19, trichlorsilan german, trichloorsilaan dutch, triclorosilano italian
PubChem CID 140487
IUPAC Name trichlorosilicon
SMILES [Si](Cl)(Cl)Cl

Specifications

Boiling Point 31°C to 32°C
Melting Point 127°C
Physical Form Liquid
Quantity 100 g
Assay Percent Range 98%
Flash Point -28°C (7°F)
Solubility Information Soluble in benzene,ether,heptane,chloroform and carbon tetrachloride.
Formula Weight 135.45
Refractive Index 1.402
Sensitivity Air and moisture sensitive
Density 1.342 g/mL
Chemical Name or Material Trichlorosilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.