accessibility menu, dialog, popup

UserName

ClSiClCl

CAS RN 10025-78-2

IUPAC Name: trichlorosilyl
Synonyms: trichlorosilane
Molecular Weight (g/mol): 134.44
Molecular Formula: Cl3Si
InChi Key: PPDADIYYMSXQJK-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)