missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylphosphane Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Tributylphosphane Oxide
Specifications
Specifications
| Chemical Name or Material | Tributylphosphane oxide |
| CAS | 814-29-9 |
| Analyte or Component Names | Tributylphosphane oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H27 O P |
| Certifications/Compliance | ISO 9001 |
| Synonym | Oseltamivir Phosphate Imp. H (EP), Oseltamivir Imp. H (EP), Oseltamivir Phosphate Impurity H, Oseltamivir Impurity H |
| InChI Formula | InChI=1S/C12H27OP/c1-4-7-10-14(13,11-8-5-2)12-9-6-3/h4-12H2,1-3H3 |
| SMILES | CCCCP(=O)(CCCC)CCCC |
| IUPAC Name | 1-dibutylphosphorylbutane |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction