missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Acetylcitrate, Mikromol™

Product Code. 17928454
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17928454

Brand: Mikromol MM0499.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tributyl Acetylcitrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tributyl acetylcitrate
CAS 77-90-7
Analyte or Component Names Tributyl acetylcitrate
Shipping Condition Room Temperature
Molecular Formula C20 H34 O8
Certifications/Compliance ISO 17025
Synonym Tributyl Acetylcitrate, 1,2,3-Propanetricarboxylic acid, 2-(acetyloxy)-, tributyl ester (9CI), Citric acid, tributyl ester, acetate (6CI,7CI,8CI), 2-Acetoxy-1,2,3-propanetricarboxylic acid tributyl ester, ATBC, Acetyl butyl citrate, Acetyl tri-n-butyl citrate, Acetyl tributyl citrate, Acetyl tris-n-butyl citrate, Acetylcitric acid tributyl ester, Blo-trol, Citroflex 4A-S, Citroflex A, Citroflex A 4, Citrofol BII, Estaflex ATC, Kanatol 3400AC, Monocizer ATBC, Morflex ATBC, NSC 3894, Pacizer 6412, Sansocizer ATBC, Scandinol SP 22, Tributyl 2-(acetyloxy)-1,2,3-propanetricarboxylic acid, Tributyl 2-acetoxy-1,2,3-propanetricarboxylate, Tributyl 2-acetyl citrate, Tributyl O-acetylcitrate, Tributyl acetylcitrate, Tributyl citrate acetate
InChI Formula InChI=1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3
SMILES CCCCOC(=O)CC(CC(=O)OCCCC)(OC(=O)C)C(=O)OCCCC
IUPAC Name tributyl 2-acetyloxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol) 402.48
Quantity 250 mg
Formula Weight 402.2254 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.