missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREX1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
189.00€
Specifications
| Antigen | TREX1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 - 1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TREX1 Polyclonal specifically detects TREX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TREX1 | |
| Western Blot 0.5 - 1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| DNA Repair, Editing and Processing Endonucleases | |
| PBS with 0.02% Sodium Azide | |
| 3'-5' exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3' repair exonuclease 1 | |
| TREX1 | |
| IgG | |
| Affinity Purified | |
| TREX1 antibody will not cross-react with the related protein TREX2. At least three isoforms of TREX1 are known to exist; this antibody will recognize all three isoforms. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_057465 | |
| 11277 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the center of human TREX1. The immunogen is located within amino acids 220 - 270 of TREX1. Amino Acid Squence: TRLYGQSPPDSHT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 31 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title