missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRERF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20062-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of TRERF1.
TREF1 encodes a zinc-finger transcriptional regulating protein which interacts with CBP/p300 to regulate the human gene CYP11A1.Specifications
| TRERF1 | |
| Polyclonal | |
| Unconjugated | |
| TRERF1 | |
| HSA277276, RAPA, RP1 139D8.5, TReP 132, TREP132, TRERF1, Zinc finger protein rapa | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 224829, 55809 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BXJ2, Q96PN7 | |
| TRERF1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction