missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (CL7111) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76531JF646
This item is not returnable.
View return policy
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Human samples. It is validated for Western Blot,KnockDown
Specifications
| TREM1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| This antibody was developed using a synthetic peptide derived from Q9NP99, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 54210 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, KnockDown | |
| CL7111 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction