missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC6A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94046-0.02ml
This item is not returnable.
View return policy
Description
TRAPPC6A Polyclonal antibody specifically detects TRAPPC6A in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TRAPPC6A | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| HSPC289, MGC2650, trafficking protein particle complex 6A, trafficking protein particle complex subunit 6A, TRAPP complex subunit 6A, TRAPPC6Adelta29-42, TRS33 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human TRAPPC6A (NP_077013.1). MADTVLFEFLHTEMVAELWAHDPDPGPGVSAGLRGEEAGATKGQKMSLSVLEGMGFRVGQALGERLPRETLAFREELDVLKFLCKDLWVAVFQKQMDSLRTNHQGTYVLQDNSFPLLLPMASGLQYLEEAPKFLAFTCGLLRGALYTLGIESVVTASVAALPVCKFQVVIPKS | |
| 0.02 mL | |
| Signal Transduction | |
| 79090 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction