missing translation for 'onlineSavingsMsg'
Learn More

TRAPPC6A Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-94046-0.1ml

 View more versions of this product

Product Code. 18630672

  • 470.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRAPPC6A Polyclonal antibody specifically detects TRAPPC6A in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRAPPC6A
Polyclonal
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
HSPC289, MGC2650, trafficking protein particle complex 6A, trafficking protein particle complex subunit 6A, TRAPP complex subunit 6A, TRAPPC6Adelta29-42, TRS33
Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human TRAPPC6A (NP_077013.1). MADTVLFEFLHTEMVAELWAHDPDPGPGVSAGLRGEEAGATKGQKMSLSVLEGMGFRVGQALGERLPRETLAFREELDVLKFLCKDLWVAVFQKQMDSLRTNHQGTYVLQDNSFPLLLPMASGLQYLEEAPKFLAFTCGLLRGALYTLGIESVVTASVAALPVCKFQVVIPKS
0.1 mL
Signal Transduction
79090
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.