Learn More
Transthyretin/Prealbumin Antibody (CL0290) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52905AF750
Product Code. 30297777
  • 770.00€ / 0.10mL
Description
Transthyretin/Prealbumin Monoclonal antibody specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transthyretin/Prealbumin
Monoclonal
Alexa Fluor 750
ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
This antibody was developed using a recombinant protein derived from P02766, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
7276
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0290
50mM Sodium Borate
Mouse
Protein A purified