Learn More
BRAND™ Blaubrand™ Eterna™ AR-GLAS™ Type 3 Class AS Measuring Pipettes
Shop All Brand Products
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
Tolerance:
±0.006 mL
±0.007 mL
±0.010 mL
±0.03 mL
±0.030 mL
±0.05 mL
±0.070 mL
±0.10 mL
Unit Size:
Pack of 12
Pack of 6
Description
- Manufactured from AR-GLAS™ Soda lime glass
- Type 3 (zero on top)
- DE-M Marking graduations
- Calibrated to deliver (TD, Ex.)
- Supplied with batch number and one batch certificate per packing unit
- USP individual certificate or DAkkS calibration certificate available upon request
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 835 |
| Material | AR-GLAS |
| Graduated | Yes |
| Graduations | 0.02 mL |
| Tolerance | ±0.070 mL |
| Volume (Metric) | 2 mL |