Learn More
VITLAB™ Transparent PFA Narrow-Mouth Bottle with Buttress Threads
Product Code. 10389590
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 10389590
Brand: VITLAB™ 109397
Only null left
Add to basket
Add to basket
VITLAB™ Transparent PFA Narrow-Mouth Bottle with Buttress Thread are supplied with a screw cap. Made of PFA, they are ideal for long-term storage of high-purity oxidants, acids, and alkalis, as well as hydrocarbons, trace analysis solvents, and standards.
  • Product Type: Storage Bottle
  • Description: Narrow Mouth with Buttress Thread
  • Material: PFA
  • Shape: Round
  • Includes: Screw Cap
Specifications
Bottle Surface Anti-adhesive and Smooth
Capacity (Metric) 100 mL
Color Transparent
Diameter (English) 1.8 in.
For Use With (Application) Saving and Storing
Includes Screw cap
Material PFA
Max. Temperature (Metric) -328°F to +500°F
Sterility Non-sterile
Temperature Range (Metric) -200°C to +260°C
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 120 mm
Product Type Narrow Mouth Bottle
Height (English) 4.7 in.
Closure Size S 28
Diameter (Metric) 45 mm
Closure Type Screw Cap