missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam trans-Zeatin, Cytokinin, Solid, Purity >98%, 10 mg
Description
- Molecular Weight: 219.24 Da
- Purity: >98%
- Form: Solid
- Description: Endogenous plant growth hormone and cytokinin. Serves as a potent AHK3 ligand (Ki = 1.3 nM).
- Biological Activity: Functions as a senescence-delaying hormone, acts as a positional cue regulating lateral root spacing, and mediates plant growth and development in vivo.
- Composition: 100% trans-Zeatin
Specifications
Specifications
| Buffer | 100% trans-Zeatin |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | AHK3 |
| Formulation | 100% trans-Zeatin |
| Quantity | 10 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?