Learn More
trans-Ethyl crotonate, 96%
Alle ansehen Thermo Scientific Chemicals Produkte
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Packungsgröße:
100g
500g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Acidity | 1mg KOH/g max. |
| Color | Colorless |
| Density | 0.9180g/mL |
| Boiling Point | 142°C to 143°C |
| Flash Point | 2°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | CH3CH=CHCO2C2H5 |
| Refractive Index | 1.4232 to 1.4272 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- trans-Ethyl crotonate
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur