Learn More
trans-2-Nonenoic acid, 98%
Product Code. 11410753
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410753 5 g 5g
11420753 25 g 25g
11430753 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11410753 Supplier Thermo Scientific Alfa Aesar Supplier No. A13228.06
Request Bulk or Custom Format
CAS: 3760-11-0 | C9H16O2 | 156.225 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3760-11-0
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00014020
InChI Key ADLXTJMPCFOTOO-BQYQJAHWSA-N
Synonym 2-nonenoic acid, nonylenic acid, trans-2-nonenoic acid, e-non-2-enoic acid, e-2-nonenoic acid, 2-nonenylic acid, 2e-nonenoic acid, 2-nonenoic acid, 2e, alpha-nonenoic acid, 2e-non-2-enoic acid
PubChem CID 5312586
IUPAC Name (E)-non-2-enoic acid
SMILES CCCCCCC=CC(=O)O
Specifications
Boiling Point 130°C to 132°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Recognizable
Quantity 5 g
UN Number UN3265
Beilstein 1721634
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material trans-2-Nonenoic acid