accessibility menu, dialog, popup

UserName

CAS RN 3760-11-0

H3COHO(Z)

CAS RN 3760-11-0

IUPAC Name: (2Z)-non-2-enoic acid
Synonyms: α-nonylenic acid
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: ADLXTJMPCFOTOO-FPLPWBNLSA-N
SMILES: CCCCCC\C=C/C(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)