Learn More
Tranexamic Acid Methyl Ester Hydrochloride, Mikromol™
Product Code. 17941636
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17941636 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17941636 Supplier Mikromol Supplier No. MM0330.090025
 Restricted Item
Estimated Shipment: 29-10-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tranexamic Acid Methyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tranexamic Acid Methyl Ester Hydrochloride
CAS 29275-88-5
Analyte or Component Names Tranexamic Acid Methyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H17 N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym Tranexamic Acid Methyl Ester Hydrochloride
InChI Formula InChI=1S/C9H17NO2.ClH/c1-12-9(11)8-4-2-7(6-10)3-5-8;/h7-8H,2-6,10H2,1H3;1H/t7-,8-;
SMILES Cl.COC(=O)[C@@H]1CC[C@@H](CN)CC1
IUPAC Name methyl 4-(aminomethyl)cyclohexane-1-carboxylate;hydrochloride
Molecular Weight (g/mol) 207.7
Quantity 25 mg
Formula Weight 207.1026 g/mol