missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAM/TICAM2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00€ - 462.00€
Specifications
| Antigen | TRAM/TICAM2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TRAM/TICAM2 Polyclonal antibody specifically detects TRAM/TICAM2 in Human samples. It is validated for Western BlotSpecifications
| TRAM/TICAM2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 353376 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC129876, MGC129877, Putative NF-kappa-B-activating protein 502, TICAM-2TIR domain-containing adapter molecule 2, TIRAP3, TIRPcytoplasmic adaptor, toll/interleukin-1 receptor (TIR) domain-containing adapter protein, Toll/interleukin-1 receptor domain-containing protein, toll-like receptor adaptor molecule 2, Toll-like receptor adaptor protein 3, TRAMNF-kappa-B-activating protein 502, TRIF-related adapter molecule, TRIF-related adaptor molecule | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-235 of human TRAM/TICAM2 (NP_067681.1). MGIGKSKINSCPLSLSWGKRHSVDTSPGYHESDSKKSEDLSLCNVAEHSNTTEGPTGKQEGAQSVEEMFEEEAEEEVFLKFVILHAEDDTDEALRVQNLLQDDFGIKPGIIFAEMPCGRQHLQNLDDAVNGSAWTILLLTENFLRDTWCNFQFYTSLMNSVNRQHKYNSVIPMRPLNNPLPRERTPFALQTINALEEESRGFPTQVERIFQESVYKTQQTIWKETRNMVQRQFIA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title