missing translation for 'onlineSavingsMsg'
Learn More

TRAK2 Monoclonal Antibody (S390-43), Invitrogen™

Product Code. 16322124
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16322124 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16322124 Supplier Invitrogen Supplier No. MA527606

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAK2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone S390-43
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Gene TRAK2
Gene Accession No. O60296, Q8R2H7
Gene Alias Trafficking kinesin-binding protein 2, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, GABA-A receptor-interacting factor 1, O-GlcNAc transferase-interacting protein of 98 kDa, EMBL AAH60681.1, Ensembl ENSMUSP00000027186 MGI 1918077, Als2cr3, Grif1, Oip98, KIAA0549, TRAK2_Human
Gene Symbols Trak2
Host Species Mouse
Immunogen Fusion protein amino acids 757-914 (C-terminus) of human TRAK2
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171086, 66008
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.