missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-25092-0.025mg
This item is not returnable.
View return policy
Description
TRAILR1/TNFRSF10A Polyclonal specifically detects TRAILR1/TNFRSF10A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRAILR1/TNFRSF10A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| Rabbit | |
| 50 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer | |
| 8797 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| AAC51226 | |
| TNFRSF10A | |
| Antibody was raised against a peptide corresponding to 19 amino acids near the carboxy terminus of human DR4 protein. The immunogen is located within the last 50 amino acids of DR4. Amino Acid Squence: DSGKFIYLEDGTGSAVSLE | |
| Delipidation and Defibrination | |
| RUO | |
| Primary | |
| Human TRAILR1/TNFRSF10A has only 1 isoform (468aa, 50kD). TRAILR1/TNFRSF10A antibody has no cross reaction to TRAILR2/TNFRSF10B. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction