Learn More
TRAFD1 Antibody (OTI3E8), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44943-30ul
This item is not returnable.
View return policy
Description
TRAFD1 Monoclonal antibody specifically detects TRAFD1 in Human samples. It is validated for Western Blot
Specifications
| TRAFD1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3E8 | |
| Western Blot 1:2000 | |
| FLN29FLN29 gene product, Protein FLN29, TRAF-type zinc finger domain containing 1, TRAF-type zinc finger domain-containing protein 1 | |
| Human recombinant protein fragment corresponding to amino acids 308-582 of human TRAFD1(NP_006691) produced in E.coli. | |
| 30 μL | |
| Immunology, Innate Immunity | |
| 10906 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |