missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRAF7 Polyclonal antibody specifically detects TRAF7 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TRAF7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | DKFZp586I021, E3 ubiquitin-protein ligase TRAF7, EC 6.3.2.-, MGC7807, RFWD1RING finger protein 119, RNF119RING finger and WD repeat-containing protein 1, TNF receptor-associated factor 7ring finger and WD repeat domain 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 260-400 of human TRAF7 (NP_115647.2). PHSKYGCTFIGNQDTYETHLETCRFEGLKEFLQQTDDRFHEMHVALAQKDQEIAFLRSMLGKLSEKIDQLEKSLELKFDVLDENQSKLSEDLMEFRRDASMLNDELSHINARLNMGILGSYDPQQIFKCKGTFVGHQGPVW |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?