missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-6 Antibody (S09-3F4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19793-50ul
This item is not returnable.
View return policy
Description
TRAF-6 Monoclonal antibody specifically detects TRAF-6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TRAF-6 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S09-3F4 | |
| Unconjugated | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
| A synthetic peptide of human TRAF-6 (Uniprot # Q9Y4K3) | |
| 50 μg | |
| Immunology, Innate Immunity, Signal Transduction, Zinc Finger | |
| 7189 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction