missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-6 Antibody (326019) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3284W
This item is not returnable.
View return policy
Description
TRAF-6 Monoclonal antibody specifically detects TRAF-6 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| TRAF-6 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 326019 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
| E.coli-derived recombinant human TRAF-6, Met1-Val522, Accession # Q9Y4K3 | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction, Zinc Finger | |
| 7189 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction