missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRAF-1 Antibody (TRAF1/3365), Novus Biologicals™

Product Code. 30043443 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30043443 20 μg 20µg
30043944 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30043443 Supplier Novus Biologicals™ Supplier No. NBP32674120ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF-1
Applications ELISA
Classification Monoclonal
Clone TRAF1/3365
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 10 mM PBS with 0.05% BSA
Gene Alias EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1
Host Species Mouse
Immunogen Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 7185
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.