Learn More
Thermo Scientific™ UHPLC Filters
Product Code. 11550944 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
UHPLC Direct Connect Filter Holder
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11550944 1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm Pack of 5
10775706 UHPLC Direct Connect Filter Holder Each
10127594 2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11550944 Supplier Thermo Scientific™ Supplier No. 22185
From Chemicals to Chromatography
Use promo code 27991
View all promotions
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Protects columns without compromising performance.
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 0.2 μm
Type 1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
Diameter (Metric) Inner 1 mm
Product Line Thermo Scientific