Learn More
Trace Element Solution 2, MP Biomedicals
Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Chemical Name or Material | Trace Element Solution 2 |
| Synonym | 1, 000X Solution |
| Quantity | 100 mL |