missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA-1-81 Mouse, anti-Human, PE, Clone: TRA-1-81, STEMCELL Technologies
Description
- The TRA-1-81 antibody reacts with a carbohydrate epitope on TRA-1-81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines
- No immunoreactivity is observed with mouse EC, EG, ES, or iPS cells
- The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase
Specifications
Specifications
| Antigen | TRA-1-81 |
| Applications | Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | TRA-1-81 |
| Conjugate | PE |
| Host Species | Mouse |
| Quantity | 25 Tests |
| Primary or Secondary | Primary |
| Target Species | Rhesus Monkey, Human, Rat |
| Isotype | IgM κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?