missing translation for 'onlineSavingsMsg'
Learn More

TRA-1-81 Mouse, anti-Human, PE, Clone: TRA-1-81, STEMCELL Technologies

Product Code. 17138811
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17138811 25 Tests 25 tests
17128811 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17138811 Supplier Stemcell Technologies Supplier No. 60065PE.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

  • The TRA-1-81 antibody reacts with a carbohydrate epitope on TRA-1-81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines
  • No immunoreactivity is observed with mouse EC, EG, ES, or iPS cells
  • The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase

Specifications

Antigen TRA-1-81
Applications Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone TRA-1-81
Conjugate PE
Host Species Mouse
Quantity 25 Tests
Primary or Secondary Primary
Target Species Rhesus Monkey, Human, Rat
Isotype IgM κ
Cell Type Pluripotent Stem Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.