missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPTE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25120-1-AP-20UL
This item is not returnable.
View return policy
Description
TPIP is a member of a large class of membrane-associated phosphatases with substrate specificity for the 3-position phosphate of inositol phospholipids.[supplied by OMIM, Jul 2002]Specifications
| TPTE2 | |
| Polyclonal | |
| Unconjugated | |
| TPTE2 | |
| Lipid phosphatase TPIP, TPIP, TPTE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 93492 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6XPS3 | |
| TPTE2 | |
| TPTE2 Fusion Protein Ag19169 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?