missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPST2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149161
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2, and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
Specifications
| TPST2 | |
| Polyclonal | |
| Unconjugated | |
| Tpst2 | |
| AI448750; D5ucla3; grm; grt; hypothetical protein LOC540183; protein-tyrosine sulfotransferase 2; Tango13b; Tpst2; TPST-2; transport and golgi organization 13 homolog B; tyrosylprotein phosphotransferase 2; tyrosylprotein sulfotransferase 2; tyrosylprotein sulfotransferase-2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 8459 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O60704 | |
| Tpst2 | |
| Chinese hamster ovary cell line CHO-derived recominant human Tyrosylprotein Sulfotransferase 2/TPST2 , Gln26-Ser377 (predicted). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction