missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10713-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.Specifications
| TPI1 | |
| Polyclonal | |
| Unconjugated | |
| TPI1 | |
| TIM, TPI, TPI1, Triose phosphate isomerase, Triosephosphate isomerase, triosephosphate isomerase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21991, 24849, 7167 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17751, P48500, P60174 | |
| TPI1 | |
| TPI1 Fusion Protein Ag1175 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction