missing translation for 'onlineSavingsMsg'
Learn More

Abcam TPCA 1, IκB Kinase Inhibitor, >98% Purity, Solid, 10 mg

Product Code. 32120556
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120556 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120556 Supplier Abcam Supplier No. ab14552210mg
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective IκB kinase inhibitor with anti-inflammatory effects.

  • Molecular Weight: 279.29 Da
  • Purity: >98%
  • Potency: Potent, selective IκB kinase inhibitor (IC50 = 17.9 nM)
  • Mechanism: Inhibits NF-κB nuclear localization
  • In vivo effects: Shows anti-inflammatory effects
  • Form: Solid
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Tpca-1
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule IκB Kinase
Formulation 100% Tpca-1
Quantity 10 mg
For Use With (Application) IkappaB kinase inhibitor, NF-kB nuclear localization inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.