missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR/mTOR Antibody (CL8228) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21165R
This item is not returnable.
View return policy
Description
TOR/mTOR Monoclonal antibody specifically detects TOR/mTOR in Human samples. It is validated for Western Blot
Specifications
| TOR/mTOR | |
| Monoclonal | |
| DyLight 550 | |
| EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR | |
| This antibody was developed using a synthetic peptide derived from P42345, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, Macroautophagy, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control | |
| 2475 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL8228 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction