missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | TOP2B |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656472
|
Novus Biologicals
NBP2-97789-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608614
|
Novus Biologicals
NBP2-97789-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOP2B Polyclonal antibody specifically detects TOP2B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| TOP2B | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.0) | |
| 7155 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen MLAA-44, DNA topoisomerase 2-beta, DNA topoisomerase II beta, DNA topoisomerase II, 180 kD, DNA topoisomerase II, beta isozyme, EC 5.99.1.3, top2beta, TOPIIB, topo II beta, topoisomerase (DNA) II beta (180kD), topoisomerase (DNA) II beta 180kDa, topoisomerase II beta, topoisomerase IIb, U937 associated antigen | |
| Produced in rabbits immunized with E. coli-derived Human TOP2B fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title