missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP1MT Mouse anti-Human, Clone: TOP1MT/613, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07562-100ug
This item is not returnable.
View return policy
Description
TOP1MT Monoclonal specifically detects TOP1MT in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| TOP1MT | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| mitochondrial, mitochondrial DNA topoisomerase I, topoisomerase (DNA) I, mitochondrial | |
| Recombinant full-length human TOP1MT protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| TOP1MT/613 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 116447 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction