missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxyaniline hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11333107
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11333107

Brand: Thermo Scientific Alfa Aesar L13490.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85006-21-9
Molecular Formula C7H9Cl2NO
Molecular Weight (g/mol) 194.06
MDL Number MFCD00012962
InChI Key NQWBPXKJBZYGHZ-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxyaniline hydrochloride, 3-amino-4-chloroanisole hydrochloride, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxyaniline hcl, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, benzenamine, 2-chloro-5-methoxy-, hydrochloride, timtec-bb sbb008038, pubchem3658, pubchem3660
PubChem CID 2723900
IUPAC Name 2-chloro-5-methoxyaniline;hydrochloride
SMILES [Cl-].COC1=CC=C(Cl)C([NH3+])=C1

Specifications

Melting Point ∽207°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3699386
Formula Weight 194.06
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-methoxyaniline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.