CAS RN 85006-21-9
CAS RN 85006-21-9
IUPAC Name:
2-chloro-5-methoxyanilinium chloride
Synonyms:
2-chloro-5-methoxyanilinium chloride
Molecular Weight (g/mol):
194.06
Molecular Formula:
C7H9Cl2NO
InChi Key:
NQWBPXKJBZYGHZ-UHFFFAOYSA-N
SMILES:
[Cl-].COC1=CC=C(Cl)C([NH3+])=C1
1
–
1
of
1
results
1
2-Chloro-5-methoxyaniline hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 85006-21-9 Molecular Formula: C7H9Cl2NO Molecular Weight (g/mol): 194.06 MDL Number: MFCD00012962 InChI Key: NQWBPXKJBZYGHZ-UHFFFAOYSA-N Synonym: 2-chloro-5-methoxyaniline hydrochloride,3-amino-4-chloroanisole hydrochloride,6-chloro-m-anisidine hydrochloride,2-chloro-5-methoxyaniline hcl,6-chloro-meta-anisidine hydrochloride,2-chloro-5-methoxyanilinehydrochloride,benzenamine, 2-chloro-5-methoxy-, hydrochloride,timtec-bb sbb008038,pubchem3658,pubchem3660 PubChem CID: 2723900 IUPAC Name: 2-chloro-5-methoxyaniline;hydrochloride SMILES: [Cl-].COC1=CC=C(Cl)C([NH3+])=C1