missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolnaftate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Tolnaftate
Specifications
Specifications
| Chemical Name or Material | Tolnaftate |
| CAS | 2398-96-1 |
| Analyte or Component Names | Tolnaftate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H17 N O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Tolnaftate, O-Naphthalen-2-yl methyl(3-methylphenyl)carbamothioate |
| InChI Formula | InChI=1S/C19H17NOS/c1-14-6-5-9-17(12-14)20(2)19(22)21-18-11-10-15-7-3-4-8-16(15)13-18/h3-13H,1-2H3 |
| SMILES | CN(C(=S)Oc1ccc2ccccc2c1)c3cccc(C)c3 |
| IUPAC Name | O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction