missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNFSF13 Recombinant Protein

Brand:  Abnova™ H00008741-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192842

  • 349.00€ / 10µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH08042
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
52.91
8
Glutathione Sepharose 4 Fast Flow
10 ug
MPASSPFLLAPKGPPGNMGGPVREPALSVALWLSWGAALGAVACAMALLTQQTELQSLRREVSRLQGTGGPSQNGEGYPWQSLPEQSSDALEAWESGERSRKRRAVLTQKQKKQHSVLHLVPINATSKDDSDVTEVMWQPALRRGRGLQAQGYGVRIQDAGVYLLYSQVLFQDVTFTMGQVVSREGQGRQETLFRCIRSMPSHPDRAYNSCYSAGVFHLHQGDILSVIIPRARAKLNLSPHGTFLGL
APRIL/CD256/TALL2/TRDL-1/UNQ383/PRO715/ligand
TNFSF13
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
8741
TNFSF13 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TNFSF13
Human
Recombinant
Solution
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers