missing translation for 'onlineSavingsMsg'
Learn More

TNF Mouse anti-Human, Alexa Fluor 700, Clone: MAB11, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 557996

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15899148

  • 545.00€ / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

The MAb11 antibody reacts with human tumor necrosis factor (TNF, also known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MAb11
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant Human TNF

Intracellular Staining

Specifications

Specifications

TNF
Monoclonal
0.2mg/mL
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Recombinant Human TNF
0.1 mg
Immunology
Human
IgG1 κ
Flow Cytometry
MAB11
Alexa Fluor 700
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.