Learn More
TNF Mouse anti-Human, Alexa Fluor 700, Clone: MAB11, BD
Mouse Monoclonal Antibody
300.00€ - 545.00€
Specifications
Antigen | TNF |
---|---|
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 700 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15810699
|
BD Biosciences
561023 |
25 μg |
300.00€
Each |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15899148
|
BD Biosciences
557996 |
0.1 mg |
545.00€
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
The MAb11 monoclonal antibody specifically binds to human tumor necrosis factor (TNF, also known as TNF- α ) protein. TNF is an efficient juxtacrine, paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN- γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The MAb11 antibody has been reported to crossreact with Rhesus Macaque TNF.
Specifications
TNF | |
Flow Cytometry | |
Alexa Fluor 700 | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Human | |
Recombinant Human TNF | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.