missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (Z74P2C5*A12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61611-0.025ml
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Fish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| TNF-alpha | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Fish | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Z74P2C5*A12 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TNF | |
| Ovalbumin-conjugated synthetic peptide | |
| 0.025 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction