missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody, Novus Biologicals™
Goat Polyclonal Antibody
287.00€ - 840.00€
Specifications
| Antigen | TNF-alpha |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602747
|
Novus Biologicals
NBP3-11332-0.025mg |
0.025 mg |
287.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632399
|
Novus Biologicals
NBP3-11332 |
0.1 mg |
840.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF-alpha Polyclonal antibody specifically detects TNF-alpha in Guinea Pig samples. It is validated for Western Blot, ELISASpecifications
| TNF-alpha | |
| Polyclonal | |
| Purified | |
| RUO | |
| Guinea Pig | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Recombinant guinea pig TNF ALPHA | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Goat | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| PBS | |
| 7124 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title