missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68039-1-IG-150UL
This item is not returnable.
View return policy
Description
The TMOD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMOD2. This antibody recognizes human, pig, rabbit, chicken antigen. The TMOD2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| TMOD2 | |
| Monoclonal | |
| 500 μg/mL | |
| MGC39481; N-TMOD; NTMOD | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Pig, Rabbit, Chicken | |
| 39-40 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1E7D1 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 29767 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction