missing translation for 'onlineSavingsMsg'
Learn More

TMEM70 Antibody (4F2), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43047

Product Code. 30279340

  • 408.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TMEM70 Monoclonal antibody specifically detects TMEM70 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMEM70
Monoclonal
Unconjugated
PBS, pH 7.3, 50% glycerol
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
ELISA, Immunohistochemistry, Western Blot
4F2
Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry 1:20 - 1:200
FLJ20533, transmembrane protein 70, transmembrane protein 70, mitochondrial
transmembrane protein 70 - (Uniprot# Q9BUB7)
100 μg
Cancer, Endocrinology, Signal Transduction
54968
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.