missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47602-25ul
This item is not returnable.
View return policy
Description
TMEM37 Polyclonal specifically detects TMEM37 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TMEM37 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Neuronal voltage-gated calcium channel gamma-like subunit, transmembrane protein 37PRPR1, voltage-dependent calcium channel gamma subunit-like protein, voltage-dependent calcium channel gamma-like subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TMEM37 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LMVSQLCEDKHSQCKWVMGS | |
| 25 μL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics | |
| 140738 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction