Learn More
TMEM119 Antibody (CL8714) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76985C
Product Code. 30307504
  • 759.00€ / 0.10mL
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEM119
Monoclonal
DyLight 650
OBIF, Osteoblast Induction Factor, TMEM119
This antibody was developed using a recombinant protein derived from Q4V9L6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL8714
50mM Sodium Borate
Mouse
Protein A purified
RUO
338773
Store at 4⪚C in the dark.