missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM119 Antibody (CL8714) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76985APC
This item is not returnable.
View return policy
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| TMEM119 | |
| Monoclonal | |
| APC | |
| OBIF, Osteoblast Induction Factor, TMEM119 | |
| This antibody was developed using a recombinant protein derived from Q4V9L6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL8714 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 338773 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction