missing translation for 'onlineSavingsMsg'
Learn More

TMED2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

188.00€ - 470.00€

Specifications

Antigen TMED2
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18634432
View Documents
Novus Biologicals
NBP2-94067-0.02ml
0.02 mL
188.00€
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18620912
View Documents
Novus Biologicals
NBP2-94067-0.1ml
0.1 mL
470.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMED2 Polyclonal antibody specifically detects TMED2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

TMED2
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Stem Cell Markers
PBS (pH 7.3), 50% glycerol
10959
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
FLJ21323, Membrane protein p24A, P24A, RNP24coated vesicle membrane protein, transmembrane emp24 domain trafficking protein 2, transmembrane emp24 domain-containing protein 2
Recombinant fusion protein containing a sequence corresponding to amino acids 60-170 of human TMED2 (NP_006806.1). EITGPDNKGIYKGDRESSGKYTFAAHMDGTYKFCFSNRMSTMTPKIVMFTIDIGEAPKGQDMETEAHQNKLEEMINELAVAMTAVKHEQEYMEVRERIHRAINDNTNSRVV
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.